Description
- Locking of door and loading hatch is dependent on temperature and pressure
- Accidental opening is not possible
- Inner vessel holder is exchangeable and easy to remove and clean
- Powerful heating elements enable rapid heating
- Rapid cooling is by water circulation and support presssure directly at external walls of media vessel using sterilized demineralized water
- Water is brought to cooling temperature using plate heat exchanger with water connection
- Total process time including heating, sterilizing and cooling to 50°C lasts from 60–120 minutes, depending on size of vessel, quantity of media and temperature of cooling water
- Support pressure avoids overboiling respectively foaming of media
- Air compressor is built-in
Specifications
16 A | |
11.6 in. | |
Preparing and sterilizing microbiological media | |
220 - 240 V (single phase), 50/60 Hz, 16 A | |
222 mm Inner Vessel | |
8.7 in. Inner Vessel | |
3.6 kw |
17 L | |
296 mm | |
MediaPrep | |
250 mm | |
9.8 in. | |
50/60 Hz | |
550 x 655 x 530 mm (21.6 x 25.7 x 20.8 in.) |