| Synonym |
6alpha-Methyl-3,20-dioxopregn-4-en-17-yl acetate, Medroxyprogesterone acetate, Megestrol Acetate Imp. A (EP), Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-methyl-, (6α)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6α-methyl-, acetate (7CI,8CI), Progesterone, 17-hydroxy-6α-methyl-, acetate (6CI), (6α)-17-(Acetyloxy)-6-methylpregn-4-ene-3,20-dione, 17α-Acetoxy-6α-methylpregn-4-ene-3,20-dione, 17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 17α-Hydroxy-6α-methylpregn-4-ene-3,20-dione acetate, 17α-Hydroxy-6α-methylprogesterone acetate, 6α-Methyl-17-acetoxyprogesterone, 6α-Methyl-17R-hydroxyprogesterone acetate, 6α-Methyl-17α-Hydroxy-4-pregnene-3,20-dione acetate, 6α-Methyl-17α-acetoxy-δ4-pregnene-3,20-dione, 6α-Methyl-17α-acetoxypregn-4-ene-3,20-dione, 6α-Methyl-17α-hydroxyprogesterone acetate, Amen, Aragest, Aragest 5, Asconale, CBP 1011, Clinofem, Clinovir, Colirest, Curretab, Cycrin, DMPA, Depo-Clinovir, Depo-Prodasone, Depo-Provera, Depo-Ralovera, Depo-SubQ Provera 104, Depo-progestin, Depo-promone, Depomedroxyprogesterone acetate, Depot-medroxyprogesterone acetate, Farlutal, Farlutin, G-Farlutal, Gestapuran, Hematrol, Hysron, Indivina, Lutopolar, Lutoral, MAP, MAP (steroid), MPA, MPA Gyn 5, Med-Pro, Medroprogesterone acetate, Medroxyprogesterone 17-acetate, Meprate, Metigestrona, NSC 21171, NSC 26386, Nadigest, Nidaxin, Novo-Medrone, Oragest, Perlutex, Perlutex Leo, Prempack C, Prodasone, Progespon, Progestalfa, Progeston, Progevera, Provera, Proverone, Ralovera, Repromap, Repromix, Sirprogen, Sodelut G, U 8839, Veramix |
| IUPAC-namn |
[(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate |