missing translation for 'onlineSavingsMsg'
Läs mer

Medroxy Progesterone, TRC

Produktkod. 30470662
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470662 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470662 Supplier Toronto Research Chemicals Supplier No. TRCM203550250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Medroxyprogesterone
CAS 520-85-4
Molekylformel C22 H32 O3
Kvantitet 250 mg
Synonym Medroxyprogesterone, 17-Hydroxy-6alpha-methylpregn-4-ene-3,20-dione, Medroxyprogesterone Acetate Imp. B (EP), Pregn-4-ene-3,20-dione, 17-hydroxy-6-methyl-, (6α)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6α-methyl- (7CI,8CI), Progesterone, 17-hydroxy-6α-methyl- (6CI), (6α)-17-Hydroxy-6-methylpregn-4-ene-3,20-dione, 17-Hydroxy-6α-methylpregn-4-ene-3,20-dione, 17-Hydroxy-6α-methylprogesterone, 17α-Hydroxy-6α-methylprogesterone, 6α-Methyl-17-hydroxyprogesterone, 6α-Methyl-17R-hydroxyprogesterone, 6α-Methyl-17α-hydroxypregn-4-ene-3,20-dione, 6α-Methyl-17α-hydroxyprogesterone, 6α-Methyl-4-pregnen-17α-ol-3,20-dione, Medoxyprogesterone, Medroxyprogesteron, NSC 27408, U 8840
InChI formel InChI=1S/C22H32O3/c1-13-11-16-17(20(3)8-5-15(24)12-19(13)20)6-9-21(4)18(16)7-10-22(21,25)14(2)23/h12-13,16-18,25H,5-11H2,1-4H3/t13-,16+,17-,18-,20+,21-,22-/m0/s1
LEDER C[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(O)C(=O)C)[C@@]4(C)CCC(=O)C=C14
IUPAC-namn (6S,8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-6,10,13-trimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 344.49
Formel vikt 344.2351
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.