Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Medroxyprogesterone-d3 |
| Förpackning | Glass Bottle |
| CAS | 162462-69-3 |
| Färg | White |
| Smältpunkt | 205°C to 210°C |
| Synonym | 17α-Hydroxy-6α-methyl-d3-4-pregnene-3,20- dione |
| Molekylvikt (g/mol) | 347.52 |
| Kvantitet | 5 mg |
| Kvalitet | Analytical Standard |