missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18710-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The myocyte enhancer factor-2 (MEF-2) family of transcription factors associated with co-repessors or co-activators to regulate development and function of T cells, neuronal cells and muscle cells. Four family members arise from alternatively spliced transcripts, termed MEF-2A, -2B, -2C and -2D. These members bind as homo- and heterodimers to the MEF-2 site in the promoter region of affected genes. Differential regulation in the expression of the four transcripts implies functional distinction for each during embryogenesis and development. The process of differentiation from mesodermal precursor cells to myoblasts has led to the discovery of a variety of tissue-specific factors that regulate muscle gene expression.Specifications
MEF2B | |
Polyclonal | |
Unconjugated | |
BORCS8-MEF2B | |
LOC729991 MEF2B, LOC729991-MEF2B, MEF2B, RSRFR2, XMEF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4207, 72368 | |
-20°C | |
Liquid |
Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D6Y4 | |
Borcs8, BORCS8-MEF2B | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |