Läs mer
Mefenacet, TRC
Produktkod. 30479147
Change view
Kvantitet:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479147 250 mg
30473639 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM205600250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mefenacet
CAS 73250-68-7
Molekylformel C16 H14 N2 O2 S
Kvantitet 250 mg
Synonym 2-(2-Benzothiazolyloxy)-N-methyl-N-phenylacetamide, BTMPA, FOE 1976, Mefenacet, NTN 801, Rancho
InChI formel InChI=1S/C16H14N2O2S/c1-18(12-7-3-2-4-8-12)15(19)11-20-16-17-13-9-5-6-10-14(13)21-16/h2-10H,11H2,1H3
LEDER CN(C(=O)COc1nc2ccccc2s1)c3ccccc3
IUPAC-namn 2-(1,3-benzothiazol-2-yloxy)-N-methyl-N-phenylacetamide
Molekylvikt (g/mol) 298.36
Formel vikt 298.0776