Läs mer
Mefenamic Acid Methyl Ester, Mikromol™
Produktkod. 30051669
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051669 25 mg
30049572 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051669 Leverantör Mikromol Leverantörsnummer MM0327.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Mefenamic Acid Methyl Ester
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mefenamic Acid Methyl Ester
CAS 1222-42-0
Analyt- eller komponentnamn Mefenamic Acid Methyl Ester
Frakt skick Room Temperature
Molekylformel C16 H17 N O2
Certifikat och efterlevnad ISO 17025
Synonym Mefenamic Acid Methyl Ester, 2-[(2,3-Dimethylphenyl)amino]-benzoic acid methyl ester, Anthranilic acid, N-2,3-xylyl-, methyl ester (7CI,8CI)
InChI formel InChI=1S/C16H17NO2/c1-11-7-6-10-14(12(11)2)17-15-9-5-4-8-13(15)16(18)19-3/h4-10,17H,1-3H3
LEDER COC(=O)c1ccccc1Nc2cccc(C)c2C
IUPAC-namn methyl 2-(2,3-dimethylanilino)benzoate
Molekylvikt (g/mol) 255.31
Kvantitet 25 mg
Formel vikt 255.1259 g/mol