Learn More
Thermo Scientific™ Megafuge 16R TX-400 Clinical Centrifuge Package
Offers exceptional capacity, ergonomics and value.
Brand:  Thermo Scientific™ 50122064
Additional Details : Weight : 100.00000kg
Product Code. 12823392
  • 103667.47 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Includes: Heraeus Megafuge 16R, 230V (Mfr PN: 75004270); TX-400 rotor, Slide coat (Mfr PN: 75003181); 400mL Round buckets (set/4) - for TX-400 rotor (Mfr PN: 75003655); Sealing caps for TX-400 bucket (set/4, includes grease) (Mfr PN: 75003656); adapters 14 x 10/15mL vac, set of 4 (Mfr PN: 75003681); Adapters 19 x 5/7mL vac, set of 4 (Mfr PN: 75003680)
Description
Description
Perfect for clinical protocols, cell culture applications and microplate processing.
Specifications
Specifications
Heraeus Megafuge 16, 230V (Mfr PN: 75004230); TX-400 rotor, Slide coat (Mfr PN: 75003181); 400mL Round buckets (set/4) - for TX-400 rotor (Mfr PN: 75003655); Sealing caps for TX-400 bucket (set/4, includes grease) (Mfr PN: 75003656); adapters 14 x 10/15mL vac, set of 4 (Mfr PN: 75003681); Adapters 19 x 5/7mL vac, set of 4 (Mfr PN: 75003680)
Megafuge 16 TX-400 Clinical Centrifuge Package
Videos