Läs mer
Megestrol, TRC
Produktkod. 30133843
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133843 500 mg
18080800 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133843 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM208040500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3562-63-8
Molekylformel C22 H30 O3
Molekylvikt (g/mol) 342.47
Synonym Pregna-4,6-diene-3,20-dione, 17-hydroxy-6-methyl-, 17-Hydroxy-6-methylpregna-4,6-diene-3,20-dione, Megestrol, 6-Methyl-17-hydroxypregna-4,6-diene-3,20-dione
IUPAC-namn (8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-6,10,13-trimethyl-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-3-one
LEDER CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Megestrol
Kvantitet 500 mg
InChI formel InChI=1S/C22H30O3/c1-13-11-16-17(20(3)8-5-15(24)12-19(13)20)6-9-21(4)18(16)7-10-22(21,25)14(2)23/h11-12,16-18,25H,5-10H2,1-4H3/t16-,17+,18+,20-,21+,22+/m1/s1
Formel vikt 342.2195