missing translation for 'onlineSavingsMsg'
Läs mer

Meglumine Diatrizoate, TRC

Produktkod. 30482065
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482065 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30482065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM208205500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Diatrizoic acid meglumine salt
CAS 131-49-7
Molekylformel C11 H9 I3 N2 O4 . C7 H17 N O5
Kvantitet 500 mg
Synonym Diatrizoic acid meglumine, D-Glucitol, 1-deoxy-1-(methylamino)-, 3,5-bis(acetylamino)-2,4,6-triiodobenzoate (1:1) (ACI), D-Glucitol, 1-deoxy-1-(methylamino)-, 3,5-bis(acetylamino)-2,4,6-triiodobenzoate (salt) (9CI), Benzoic acid, 3,5-bis(acetylamino)-2,4,6-triiodo-, compd. with 1-deoxy-1-(methylamino)-D-glucitol (1:1) (9CI), Benzoic acid, 3,5-diacetamido-2,4,6-triiodo-, compd. with 1-deoxy-1-(methylamino)glucitol (7CI), Benzoic acid, 3,5-diacetamido-2,4,6-triiodo-, compd. with 1-deoxy-1-methylaminosorbitol (6CI), Glucitol, 1-deoxy-1-(methylamino)-, 3,5-diacetamido-2,4,6-triiodobenzoate (salt), D- (8CI), Sorbitol, 1-deoxy-1-methylamino-, compd. with 3,5-diacetamido-2,4,6-triiodobenzoic acid (6CI), 3,5-Diacetamido-2,4,6-triiodobenzoic acid methylglucamine salt, Amidotrizoic acid methylglucamine salt, Angiografin, Angiografin 310, Angiografin 60, Angiografin 65, Angiographin, Angiovist 282, Cardiografin, Diatrizoate meglumine, Diatrizoate methylglucamine, Ditrizoate methylglucamine, Gastrografin, Hypaque 13.4, Hypaque 60, Hypaque M 30, Hypaque meglumine, Meglumine amidotrizoate, Meglumine diatrizoate, Methylglucamine 3,5-diacetamido-2,4,6-triiodobenzoate, Methylglucamine diacetyldiaminetriiodobenzoate, Methylglucamine diatrixoate, Methylglucamine diatrizoate, Methylglucammonium 2,4,6-triiodo-3,5-dipropionamidobenzoate, N-Methylglucamine diatrizoate, Reno M 60, Reno-M-Dip, Renograffin M 76, Renografin, Renurix, Unipaque, Urografic acid methylglucamine salt, Urovist
InChI formel InChI=1S/C11H9I3N2O4.C7H17NO5/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18;1-8-2-4(10)6(12)7(13)5(11)3-9/h1-2H3,(H,15,17)(H,16,18)(H,19,20);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1
LEDER CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC(=O)Nc1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I
IUPAC-namn 3,5-diacetamido-2,4,6-triiodobenzoic acid;(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
Molekylvikt (g/mol) 809.13
Formel vikt 808.8803
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.