missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | MEIS3 |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850674
|
Proteintech
12775-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840674
|
Proteintech
12775-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Transcriptional regulator which directly modulates PDPK1 expression, thus promoting survival of pancreatic beta-cells. Also regulates expression of NDFIP1, BNIP3, and CCNG1.Specifications
MEIS3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P97368, Q99687 | |
17537, 361514, 56917 | |
MEIS3 Fusion Protein Ag3471 | |
Primary | |
-20°C | |
MEIS3 |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Homeobox protein Meis3, Meis homeobox 3, Meis1 related protein 2, MEIS3, MRG2 | |
Meis3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |