missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MEK1 Antibody (OTI1F5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72643
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MEK1 Monoclonal specifically detects MEK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifikationer
| MEK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) | |
| Mouse | |
| 43.4 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1F5 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| MAP2K1 | |
| Full-length protein expressed in 293T cell transfected with human MAP2K1 expression vector | |
| Affinity Purified | |
| Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase, Signal Transduction, Tyrosine Kinases | |
| 5604 | |
| Human, Mouse, Rat | |
| Lyophilized |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering