Läs mer
Melamine-13C3,15N3, TRC
Produktkod. 30012800
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012800 10 mg
30005844 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012800 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM20870410MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246816-14-7
Molekylformel 13C3 H6 N3 15N3
Molekylvikt (g/mol) 132.08
Synonym 1,3,5-Triazine-2,4,6-triamine-13C3,15N3, 2,4,6-Triamino-s-triazine-13C3,15N3, Cyanurotriamide-13C3,15N3, NSC 2130-13C3,15N3;
IUPAC-namn 1,3,5-triazine-2,4,6-13C3-2,4,6-triamine-15N3
LEDER [15NH2][13C]1=N[13C]([15NH2])=N[13C]([15NH2])=N1
Specifikationer
Kemiskt namn eller material Melamine-13C3,15N3
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 132.07
Procent renhet >95%