Läs mer
Melamine-15N3, TRC
Produktkod. 30006590
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006590 10 mg
30011172 1 mg
30004050 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006590 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM20870310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 287476-11-3
Molekylformel C3 H6 15N3 N3
Molekylvikt (g/mol) 129.1
Synonym Melamine triamine 15N3, Melamine Triamine-15N3, 1,3,5-Triazine-2,4,6-tri(amine-15N) (9CI, ACI)
IUPAC-namn 1,3,5-triazine-2,4,6-triamine
LEDER [15NH2]c1nc([15NH2])nc([15NH2])n1
Specifikationer
Kemiskt namn eller material Melamine-15N3
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)/i4+1,5+1,6+1
Formel vikt 129.0565
Procent renhet >95%