Läs mer
Melamine Cyanurate, TRC
Produktkod. 30132769
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132769 100 mg
30004902 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132769 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM208720100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 37640-57-6
Molekylformel C3 H6 N6 . C3 H3 N3 O3
Molekylvikt (g/mol) 255.19
Synonym 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, compd. with 1,3,5-triazine-2,4,6-triamine (1:1) (9CI, ACI), 1,3,5-Triazine-2,4,6-triamine, compd. with 1,3,5-triazine-2,4,6(1H,3H,5H)-trione (1:1) (9CI), Addiflam POW-MCA, Aflammit PMN 525, Budit 314, Budit 315, Budit 315E, C 1, CG 610, CG 615, D 022, DHF 100, Exflam MCA, FCC-MC 03, Flameproof MC-M, FR 106, FR 6120, FT 6120MC15, Fyrol MC, GA 3437, Haida FR 106, HT 211, JLS-MC, JLS-MC 25, JLS-MC 810, JLS-MC 810D, JLS-MCA, JSL-MC 15, Mastertek 374115, MC 1, MC 1 (cyanurate), MC 101, MC 101 (cyanurate), MC 105, MC 105 (cyanurate), MC 1100, MC 12, MC 15, MC 1N, MC 2010N, MC 20S, MC 20SJ, MC 25, MC 3000, MC 4000, MC 410, MC 4500, MC 490, MC 50, MC 5F, MC 5S, MC 600, MC 6000, MC 602, MC 610, MC 640, MC 690, MC 810, MC 810 (fireproofing agent), MC White, MC-C 0, MCA, MCA (fireproofing agent), MCA (isocyanurate), MCA 01, MCA 033, MCA 10, MCA 1000, MCA 12, MCA 15, MCA 151, MCA 152, MCA 19, MCA 210, MCA 25, MCA 253, MCA 300, MCA 301, MCA 410, MCA 50, MCA 50 (cyanurate), MCA 510, MCA 55, MCA 55 (cyanurate), MCA-A, MCA-C 0, MCA-C 1, MCA-CO, MCA-COP, Melagard MC 25, Melagard MC 8, Melamine cyanurate, Melamine cyanurate (1:1), Melamine isocyanurate, Melamine isocyanurate (1:1), Melamine-cyanuric acid compd., Melamine-isocyanuric acid compd., Melanic MC, Melapur 25, Melapur 50, Melapur MC, Melapur MC 15, Melapur MC 15ED, Melapur MC 25, Melapur MC 50, Mitec MX 601, MP, MX, MX 44, MX 601, NC 600, Nonfla MC 600, Nord-Min MC 25J, NSC 231587, Phoslite B 65AM, Plastisan B, Rilsan MB 3000, Stabiace MC 2010N, Stabiace MC 3S, Stabiace MC 5F, Stabiace MC 5S, SunFla MC 100, XS-MC 1, XS-MC 15, XS-MC 151
IUPAC-namn 1,3,5-triazinane-2,4,6-trione;1,3,5-triazine-2,4,6-triamine
LEDER Nc1nc(N)nc(N)n1.O=C2NC(=O)NC(=O)N2
Specifikationer
Kemiskt namn eller material Melamine Cyanurate
Kvantitet 100 mg
InChI formel InChI=1S/C3H6N6.C3H3N3O3/c4-1-7-2(5)9-3(6)8-1;7-1-4-2(8)6-3(9)5-1/h(H6,4,5,6,7,8,9);(H3,4,5,6,7,8,9)
Formel vikt 255.0828