missing translation for 'onlineSavingsMsg'
Learn More

Melanophilin Mouse anti-Human, Porcine, Clone: 1A8D12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66092-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827655

  • 4721.67 SEK / 150µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Slac2-a (for synaptotagmin-like protein (Slp) homolog lacking C2 domains-a) links Rab27A on melanosomes with Myosin VA in melanocytes. Slac2-a binds Myosin VA through a C-terminal region and GTP-bound Rab27A through its synaptotagmin-like protein homology domain (SHD), which is located near the N-terminus. The transport of pigment and cytotoxic granules in melanocytes requires the stable formation of this complex, and thus mutations in the binding domains of the three protein components may cause albinism and/or severe immune disorders.
Specifications

Specifications

Melanophilin
Monoclonal
0.77 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BV36
MLPH
Melanophilin Fusion Protein Ag19286
150 μL
Primary
Human, Porcine
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1A8D12
Unconjugated
MLPH
Exophilin 3, melanophilin, MLPH, SLAC2 A, SLAC2A, Synaptotagmin like protein 2a
Mouse
Protein G
RUO
100049680, 79083
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers