Läs mer
Melengestrol Acetate, TRC
Produktkod. 30136723
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136723 25 mg
30132550 100 mg
18003010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136723 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21535025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2919-66-6
Molekylformel C25 H32 O4
Molekylvikt (g/mol) 396.52
Synonym Pregna-4,6-diene-3,20-dione, 17-(acetyloxy)-6-methyl-16-methylene-, Pregna-4,6-diene-3,20-dione, 17-hydroxy-6-methyl-16-methylene-, acetate (7CI,8CI), 17-(Acetyloxy)-6-methyl-16-methylenepregna-4,6-diene-3,20-dione, 17-Acetoxy-6-methyl-16-methylenepregna-4,6-diene-3,20-dione, 17-Hydroxy-6-methyl-16-methylenepregna-4,6-diene-3,20-dione acetate, 17α-Acetoxy-6-methyl-16-methylenepregna-4,6-diene-3,20-dione, 5373, 6-Dehydro-16-methylene-17α-hydroxy-6α-methylprogesterone acetate, 6-Methyl-16-methylene-17α-hydroxy-4,6-pregnadiene-3,20-dione 17-acetate, BDH 1921, Heifermax 500, MGA, MGA 100, MGA 100 (steroid), MGA 264, Melengestrol acetate, NSC-70968
IUPAC-namn [(8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-16-methylidene-3-oxo-1,2,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-17-yl] acetate
LEDER CC(=O)O[C@]1(C(=O)C)C(=C)C[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Melengestrol acetate
Kvantitet 25 mg
InChI formel InChI=1S/C25H32O4/c1-14-11-19-20(23(5)9-7-18(28)13-21(14)23)8-10-24(6)22(19)12-15(2)25(24,16(3)26)29-17(4)27/h11,13,19-20,22H,2,7-10,12H2,1,3-6H3/t19-,20+,22+,23-,24+,25+/m1/s1
Formel vikt 396.2301