Läs mer
Melengestrol, TRC
Produktkod. 30012830
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30012830 10 mg -
30006419 5 mg -
30005614 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21534010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5633-18-1
Molekylformel C23 H30 O3
Molekylvikt (g/mol) 354.48
Synonym Pregna-4,6-diene-3,20-dione, 17-hydroxy-6-methyl-16-methylene-, 17-Hydroxy-6-methyl-16-methylenepregna-4,6-diene-3,20-dione, Melengesterol, Melengestrel, Melengestrol
IUPAC-namn (8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-6,10,13-trimethyl-16-methylidene-1,2,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-3-one
LEDER CC(=O)[C@@]1(O)C(=C)C[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Melengestrol
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C23H30O3/c1-13-10-17-18(21(4)8-6-16(25)12-19(13)21)7-9-22(5)20(17)11-14(2)23(22,26)15(3)24/h10,12,17-18,20,26H,2,6-9,11H2,1,3-5H3/t17-,18+,20+,21-,22+,23+/m1/s1
Formel vikt 354.2195
Procent renhet >95%