Läs mer
Melphalan, 95 %, MP Biomedicals™
Produktkod. 11437632 Handla allt MP Biomedicals Produkter
Change view
Kvantitet:
100 mg
250 mg
1 g
Förpackningsstorlek:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11437632 100 mg 100mg
11447632 250 mg 250mg
11457632 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11437632 Leverantör MP Biomedicals Leverantörsnummer 0215534580
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Kemiska identifierare
CAS 148-82-3
Molekylformel C13H18Cl2N2O2
Molekylvikt (g/mol) 305.199
InChI-nyckel SGDBTWWWUNNDEQ-LBPRGKRZSA-N
Synonym melphalan, alkeran, melfalan, phenylalanine mustard, l-sarcolysine, l-pam, l-sarcolysin, l-phenylalanine mustard, levofalan, p-l-sarcolysin
PubChem CID 460612
ChEBI CHEBI:28876
IUPAC-namn (2S)-2-amino-3-[4-[bis(2-kloretyl)amino]fenyl]propansyra
LEDER C1=CC(=CC=C1CC(C(=O)O)N)N(CCCl)CCCl
Specifikationer
Kemiskt namn eller material Melphalan
Färg Gult
Analysprocentintervall 95%
Kvantitet 100 mg
Formel vikt 305.2
Fysisk form Pulver