Läs mer
Melphalan Dimer Hydrochloride, TRC
Produktkod. 30475525
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475525 5 mg
30481822 1 mg
30474732 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2169055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2238824-44-5
Molekylformel C26H35Cl3N4O4 . (HCl) x
Molekylvikt (g/mol) 610.4
Synonym 4-[[2-[[4-[Bis(2-chloroethyl)amino]-L-phenylalanyl]oxy]ethyl](2-chloroethyl)amino]-L-phenylalanine Hydrochloride
IUPAC-namn (S)-2-amino-3-(4-((2-(((S)-2-amino-3-(4-(bis(2-chloroethyl)amino)phenyl)propanoyl)oxy)ethyl)(2-chloroethyl)amino)phenyl)propanoic acid hydrochloride
LEDER ClCCN(C1=CC=C(C=C1)C[C@@H](C(OCCN(CCCl)C2=CC=C(C=C2)C[C@@H](C(O)=O)N)=O)N)CCCl.Cl
Specifikationer
Kemiskt namn eller material Melphalan Dimer Hydrochloride
Kvantitet 5 mg
InChI formel InChI=1S/C26H35Cl3N4O4.ClH/c27-9-12-32(13-10-28)21-5-3-20(4-6-21)18-24(31)26(36)37-16-15-33(14-11-29)22-7-1-19(2-8-22)17-23(30)25(34)35;/h1-8,23-24H,9-18,30-31H2,(H,34,35);1H/t23-,24-;/m0./s1