Läs mer
Melphalan Methyl Ester Hydrochloride, TRC
Produktkod. 30472958
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30472958 10 mg
1 options
Produktkod. 30472958 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21693010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Melphalan Methyl Ester Hydrochloride
CAS 62978-52-3
Molekylformel C14H21Cl3N2O2
Kvantitet 10 mg
Synonym 4-[Bis(2-chloroethyl)amino]-L-phenylalanine Methyl Ester Hydrochloride, 4-[Bis(2-chloroethyl)amino]-L-phenylalanine Methyl Ester Monohydrochloride, NSC 133726
InChI formel InChI=1S/C14H20Cl2N2O2.ClH/c1-20-14(19)13(17)10-11-2-4-12(5-3-11)18(8-6-15)9-7-16;/h2-5,13H,6-10,17H2,1H3;1H/t13-;/m0./s1
LEDER Cl.COC(=O)[C@@H](N)Cc1ccc(cc1)N(CCCl)CCCl
IUPAC-namn methyl (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoate;hydrochloride
Molekylvikt (g/mol) 355.69
Formel vikt 354.0669