Läs mer
Memantine, TRC
Produktkod. 30480724
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480724 25 mg
30478569 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30480724 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21799525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Memantine
CAS 19982-08-2
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H21 N
Kvantitet 25 mg
Synonym Tricyclo[3.3.1.13,7]decan-1-amine, 3,5-dimethyl-, 1-Adamantanamine, 3,5-dimethyl- (8CI), 3,5-Dimethyltricyclo[3.3.1.13,7]decan-1-amine, 1,3-Dimethyl-5-adamantanamine, 1,3-Dimethyl-5-aminoadamantane, 1-Amino-3,5-dimethyladamantane, 3,5-Dimethyl-1-adamantanamine, 3,5-Dimethyl-1-adamantylamine, Alzantin, D 145, D 145 (sympathomimetic), DMAA, DRG 0267, Ebixa, Memantina, Memantine
InChI formel InChI=1S/C12H21N/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10/h9H,3-8,13H2,1-2H3
LEDER CC12CC3CC(C)(C1)CC(N)(C3)C2
IUPAC-namn 3,5-dimethyladamantan-1-amine
Molekylvikt (g/mol) 179.3
Formel vikt 179.1674