Learn More
Thermo Scientific™ Membrane Kit for Dissolved Oxygen Probe
Includes six membrane caps, polishing disk and electrolyte filling solution. Thermo Scientific™ Meembrane Kit for Dissolved Oxygen Probe is for use with Accumet™ Benchtop Dissolved Oxygen Meters.
Brand:  Thermo Scientific™ 13637DOM
Additional Details : Weight : 0.99000kg
Product Code. 10319657
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • For use with Accumet™ Benchtop Dissolved Oxygen Meter
  • Includes 6 membrane caps, polishing disk, electrolyte filling solution
Specifications
Specifications
Accumet™ Benchtop Dissolved Oxygen Meter
6 membrane caps, polishing disk, electrolyte filling solution