Learn More
Thermo Scientific™ Membrane Kit for Dissolved Oxygen Probe
Includes six membrane caps, polishing disk and electrolyte filling solution. Thermo Scientific™ Meembrane Kit for Dissolved Oxygen Probe is for use with Accumet™ Benchtop Dissolved Oxygen Meters.
Brand: Thermo Scientific™ 13637DOM
Additional Details : Weight : 0.99000kg
Description
- For use with Accumet™ Benchtop Dissolved Oxygen Meter
- Includes 6 membrane caps, polishing disk, electrolyte filling solution
Specifications
Accumet™ Benchtop Dissolved Oxygen Meter | |
6 membrane caps, polishing disk, electrolyte filling solution |