missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Menin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 26 publications
Brand: Novus Biologicals NB100-215
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Menin Polyclonal specifically detects Menin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifikationer
| Menin | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| menin, multiple, multiple endocrine neoplasia I | |
| Rabbit | |
| 68 kDa | |
| 100 μL | |
| Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors | |
| 4221 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:10000, Immunohistochemistry, Immunoprecipitation 2-10 ug/mg of lysate, Immunohistochemistry-Paraffin 1:100-1:250 | |
| O00255 | |
| MEN1 | |
| The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 615) of human Multiple Endocrine Neoplasia 1 using the numbering given in entry NP_000235.2 (GeneID 4221). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering