Läs mer
(±)-Menthone, TRC
Produktkod. 30464778
Change view
Kvantitet:
100 mg
1 g
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464778 2 g
30482303 100 mg
30479733 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464778 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2191352G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 89-80-5
Molekylformel C10H18O
Molekylvikt (g/mol) 154.25
Synonym trans-5-Methyl-2-(1-methylethyl)cyclohexanone, trans-p-Menthan-3-one, DL-Menthone, Menthone, Menthone G, Neomenthone, dl-Menthone, p-Menthone, trans-Menthan-3-one, trans-Menthone, trans-p-Menthan-3-one, trans-p-Menthone
LEDER OC[C@H](O)CO[C@H]1C(O)C(O)[C@@H](O)C(CO)O1
Specifikationer
Kemiskt namn eller material (±)-Menthone
Anteckningar om renhetsgrad GC
Kvantitet 2 g
InChI formel InChI=1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9+/m1/s1
Procent renhet >95%