Läs mer
(+)-Menthone, TRC
Produktkod. 30137614
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137614 10 g
30007659 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137614 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM19955510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3391-87-5
Molekylformel C10H18O
Molekylvikt (g/mol) 154.25
Synonym (+)-Menthone, (2R,5S)-5-Methyl-2-(1-methylethyl)cyclohexanone, D-Menthone, (2R-trans)-5-Methyl-2-(1-methylethyl)-cyclohexanone, (1S,4R)-(+)-p-Menthan-3-one
IUPAC-namn (2R,5S)-2-isopropyl-5-methylcyclohexan-1-one
LEDER O=C1[C@@H](C(C)C)CC[C@H](C)C1
Specifikationer
Kemiskt namn eller material (+)-Menthone
Kvantitet 10 g
InChI formel InChI=1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9+/m0/s1
Formel vikt 154.14