Läs mer
Menthyl Acetate, TRC
Produktkod. 30478355
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30478355 500 mg
1 options
Produktkod. 30478355 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM218716500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Menthyl Acetate
CAS 16409-45-3
Molekylformel C12 H22 O2
Kvantitet 500 mg
Synonym Menthyl Acetate, Cyclohexanol, 5-methyl-2-(1-methylethyl)-, 1-acetate (ACI), Cyclohexanol, 5-methyl-2-(1-methylethyl)-, acetate (9CI), Menthol, acetate (6CI, 8CI), 2-Isopropyl-5-methylcyclohexyl acetate, Acetic acid 5-methyl-2-(1-methylethyl)cyclohexyl es
InChI formel InChI=1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3
LEDER CC(C)C1CCC(C)CC1OC(=O)C
IUPAC-namn (5-methyl-2-propan-2-ylcyclohexyl) acetate
Molekylvikt (g/mol) 198.302
Formel vikt 198.162