Läs mer
(±)-Menthyl Acetate, TRC
Produktkod. 30482683
Change view
Kvantitet:
10 mL
Produktkod. Kvantitet
30482683 10 mL
1 options
Produktkod. 30482683 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21871510ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (±)-Menthyl Acetate
CAS 89-48-5
Anteckningar om renhetsgrad GC
Molekylformel C12 H22 O2
Kvantitet 10 mL
Synonym (±)-Menthol Acetate, Menthyl Acetate, dl-Menthyl Actate, cis-1,3,trans-1,4-Menthol Acetate, (1R,2S,5R)-rel-5-Methyl-2-(1-methylethyl)cyclohexyl Acetate, (1R,2S,5R)-rel-2-Isopropyl-5-methylcyclohexyl Acetate
InChI formel InChI=1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3/t9-,11+,12-/m1/s1
LEDER CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)C
IUPAC-namn [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] acetate
Molekylvikt (g/mol) 198.302
Formel vikt 198.162