Läs mer
Mepanipyrim-2-hydroxypropyl, TRC
Produktkod. 30133606
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133606 100 mg
30003539 10 mg
30008780 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133606 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM223605100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 204571-52-8
Molekylformel C14 H17 N3 O
Molekylvikt (g/mol) 243.3
Synonym Mepanipyrim-2-hydroxypropyl, α,6-Dimethyl-2-(phenylamino)-4-pyrimidineethanol
IUPAC-namn 1-(2-anilino-6-methylpyrimidin-4-yl)propan-2-ol
LEDER CC(O)Cc1cc(C)nc(Nc2ccccc2)n1
Specifikationer
Kemiskt namn eller material Mepanipyrim-2-hydroxypropyl
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C14H17N3O/c1-10-8-13(9-11(2)18)17-14(15-10)16-12-6-4-3-5-7-12/h3-8,11,18H,9H2,1-2H3,(H,15,16,17)
Formel vikt 243.1372
Procent renhet >95%