Läs mer
Meperfluthrin, TRC
Produktkod. 30137704
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137704 25 mg
30133552 100 mg
17953825 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137704 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22380025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 915288-13-0
Molekylformel C17 H16 Cl2 F4 O3
Molekylvikt (g/mol) 415.21
Synonym Meperfluthrin, [2,3,5,6-Tetrafluoro-4-(methoxymethyl)phenyl]methyl (1R,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate, 2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl (1R,3S)-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, 2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl (1R)-trans-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
IUPAC-namn [2,3,5,6-tetrafluoro-4-(methoxymethyl)phenyl]methyl (1R,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
LEDER COCc1c(F)c(F)c(COC(=O)[C@@H]2[C@@H](C=C(Cl)Cl)C2(C)C)c(F)c1F
Specifikationer
Kemiskt namn eller material Meperfluthrin
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C17H16Cl2F4O3/c1-17(2)9(4-10(18)19)11(17)16(24)26-6-8-14(22)12(20)7(5-25-3)13(21)15(8)23/h4,9,11H,5-6H2,1-3H3/t9-,11+/m1/s1
Formel vikt 414.0413
Procent renhet >95%