Läs mer
Mephenoxalone, TRC
Produktkod. 30471660
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471660 25 mg
30472876 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30471660 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22490025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70-07-5
Molekylformel C11 H13 N O4
Molekylvikt (g/mol) 223.23
Synonym 2-Oxazolidinone, 5-[(2-methoxyphenoxy)methyl]-, 2-Oxazolidinone, 5-[(o-methoxyphenoxy)methyl]- (6CI,7CI,8CI), 5-[(2-Methoxyphenoxy)methyl]-2-oxazolidinone, (±)-Mephenoxalone, 5-(o-Methoxyphenoxymethyl)-2-oxazolidinone, 5-(o-Methoxyphenoxymethyl)-2-oxazolidone, AHR 233, Control-OM, Dorsiflex, Dorsilon, Ekilan, Lenetran, Lenetran Tab, Lenetranat, Mephenoxalone, Methoxadone, Methoxydon, Methoxydone, Metoxadon, Metoxadone, Moderamin, OM-518, Placidex, Riself, Tranpoise, Trepidone, Valans, Xerene
IUPAC-namn 5-[(2-methoxyphenoxy)methyl]-1,3-oxazolidin-2-one
LEDER COc1ccccc1OCC2CNC(=O)O2
Specifikationer
Kemiskt namn eller material Mephenoxalone
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C11H13NO4/c1-14-9-4-2-3-5-10(9)15-7-8-6-12-11(13)16-8/h2-5,8H,6-7H2,1H3,(H,12,13)
Formel vikt 223.0845
Procent renhet >95%