Läs mer
Mephentermine Hemisulphate, Mikromol™
Produktkod. 30049067
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30049067 500 mg
1 options
Produktkod. 30049067 Leverantör Mikromol Leverantörsnummer MM0740.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Mephentermine Hemisulphate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mephentermine Hemisulphate
CAS 1212-72-2
Analyt- eller komponentnamn Mephentermine Hemisulphate
Frakt skick Room Temperature
Molekylformel 2 C11 H17 N . H2 O4 S
Certifikat och efterlevnad ISO 17034
Synonym Mephentermine Hemisulphate, 2-Methylamino-2-methyl-1-phenylpropane sulfate, Mephentermine sulfate, Mephentermine sulphate, Wyamine sulfate, Benzeneethanamine, N,alpha,alpha-trimethyl-, sulfate (2:1), Phenethylamine, N,alpha,alpha-trimethyl-, sulfate (2:1) (8CI), N,alpha,alpha-Trimethylphenethylamine Sulfate
InChI formel InChI=1S/2C11H17N.H2O4S/c2*1-11(2,12-3)9-10-7-5-4-6-8-10;1-5(2,3)4/h2*4-8,12H,9H2,1-3H3;(H2,1,2,3,4)
LEDER CNC(C)(C)Cc1ccccc1.CNC(C)(C)Cc2ccccc2.OS(=O)(=O)O
IUPAC-namn N,2-dimethyl-1-phenylpropan-2-amine;sulfuric acid
Molekylvikt (g/mol) 424.6
Kvantitet 500 mg
Formel vikt 424.2396 g/mol