Läs mer
Mepivacaine hydrochloride, MedChemExpress
Produktkod. p-200099072
Encompass_Preferred
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16490968 5 g
16452209 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16490968 Leverantör Medchem Express Leverantörsnummer HYB0517A
Lägg i varukorgen
Lägg i varukorgen
MedChemExpress Mepivacaine hydrochloride binds to specific voltage-gated sodium ion channels in neuronal cell membranes, which inhibits both sodium influx and membrane depolarization.
Mepivacaine hydrochloride binds to specific voltage-gated sodium ion channels in neuronal cell membranes, which inhibits both sodium influx and membrane depolarization.
Specifikationer
Kemiskt namn eller material Mepivacaine hydrochloride
CAS 1722-62-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C15H23ClN2O
Kvantitet 5 g
Löslighetsinformation H2O : 100 mg/mL (353.59 mM; Need ultrasonic)
LEDER O=C(C(CCCC1)N1C)NC(C(C)=CC=C2)=C2C.Cl
Molekylvikt (g/mol) 282.81
Formel vikt 282.81
Procent renhet 99.83%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation