Läs mer
Mepivacaine N-Oxide, TRC
Produktkod. 30135905
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135905 250 mg
30040369 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135905 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM225065250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346597-75-8
Molekylformel C15 H22 N2 O2
Molekylvikt (g/mol) 262.35
Synonym 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-1-methyl-, 1-oxide, Mepivacaine N-oxide, N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide 1-oxide
IUPAC-namn N-(2,6-dimethylphenyl)-1-methyl-1-oxidopiperidin-1-ium-2-carboxamide
LEDER Cc1cccc(C)c1NC(=O)C2CCCC[N+]2(C)[O-]
Specifikationer
Kemiskt namn eller material Mepivacaine N-Oxide
Kvantitet 250 mg
InChI formel InChI=1S/C15H22N2O2/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13,3)19/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18)
Formel vikt 262.1681