Läs mer
Meprylcaine Hydrochloride, TRC
Produktkod. 30481931
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30481931 50 mg
1 options
Produktkod. 30481931 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22498050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Meprylcaine HCl
CAS 956-03-6
Molekylformel C14 H21 N O2 . Cl H
Kvantitet 50 mg
Synonym 2-Methyl-2-(propylamino)-1-propanol Benzoate (Ester) Hydrochloride, 2-Methyl-2-(propylamino)-1-propanol Benzoate Hydrochloride, 2-Methyl-2-propylaminopropyl Benzoate Hydrochloride, Meprylcaine Hydrochloride, Oracaine Hydrochloride
InChI formel InChI=1S/C14H21NO2.ClH/c1-4-10-15-14(2,3)11-17-13(16)12-8-6-5-7-9-12;/h5-9,15H,4,10-11H2,1-3H3;1H
LEDER Cl.CCCNC(C)(C)COC(=O)c1ccccc1
IUPAC-namn [2-methyl-2-(propylamino)propyl] benzoate;hydrochloride
Molekylvikt (g/mol) 271.78
Formel vikt 271.1339