Learn More
Trajan Scientific and Medical™ MEPS BINs for XCHANGE/eVol Syringes
Miniaturization of conventional sample preparation devices. Trajan Scientific and Medical MEPS BINs for XCHANGE/eVol Syringes are micro extraction by packed sorbent.
Brand: Trajan Scientific and Medical™ 2900702
Additional Details : Weight : 0.01000kg
Description
- Save hours in sample preparation
- Reduce solvent consumption and sample volume
- Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifications
MEPS Barrel Insert and Needle (BIN) | |
C8 | |
XCHANGE/eVol Syringes | |
LC Tipped Needle |