Learn More
Trajan Scientific and Medical™ MEPS BINs for XCHANGE/eVol Syringes
Miniaturization of conventional sample preparation devices. Trajan Scientific and Medical MEPS BINs for XCHANGE/eVol Syringes are micro extraction by packed sorbent.
Brand:  Trajan Scientific and Medical™ 2900702
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15269243
  • 3690.00 SEK / Pack of 5
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifications
Specifications
MEPS Barrel Insert and Needle (BIN)
C8
XCHANGE/eVol Syringes
LC Tipped Needle
Videos