Learn More
Trajan Scientific and Medical™ MEPS BINs for XCHANGE/eVol Syringes
Miniaturization of conventional sample preparation devices. Trajan Scientific and Medical MEPS BINs for XCHANGE/eVol Syringes are micro extraction by packed sorbent.
Brand:  Trajan Scientific and Medical™ 2900712
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15229253
  • 3690.00 SEK / Pack of 5
Estimated Shipment
08-12-2020
Add to basket
Description
Description
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifications
Specifications
XCHANGE/eVol Syringes
C8
MEPS Barrel Insert and Needle (BIN)
Cone Tipped Needle