Läs mer
Meptazinol Hydrochloride, TRC
Produktkod. 30466563
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30466563 10 mg
1 options
Produktkod. 30466563 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22508010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Meptazinol Hydrochloride
CAS 59263-76-2
Molekylformel C15 H23 N O . Cl H
Kvantitet 10 mg
Synonym Phenol, 3-(3-ethylhexahydro-1-methyl-1H-azepin-3-yl)-, hydrochloride (1:1), Phenol, 3-(3-ethylhexahydro-1-methyl-1H-azepin-3-yl)-, hydrochloride (9CI), IL 22811 HCl, Meptamizol hydrochloride, Meptazinol hydrochloride, Meptid, Wy 22811, Wy 22811 HCl
InChI formel InChI=1S/C15H23NO.ClH/c1-3-15(9-4-5-10-16(2)12-15)13-7-6-8-14(17)11-13;/h6-8,11,17H,3-5,9-10,12H2,1-2H3;1H
LEDER Cl.CCC1(CCCCN(C)C1)c2cccc(O)c2
IUPAC-namn 3-(3-ethyl-1-methylazepan-3-yl)phenol;hydrochloride
Molekylvikt (g/mol) 269.81
Formel vikt 269.1546