Läs mer
Meptyldinocap, MedChemExpress
Produktkod. 16443407
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16443407 50 mg
16427378 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16443407 Leverantör Medchem Express Leverantörsnummer HY17522
MedChemExpress Meptyldinocap (2,4-DNOPC) is a novel powdery mildew (Erysiphe necator) fungicide which shows protectant and post-infective activities.
Meptyldinocap (2,4-DNOPC) is a novel powdery mildew (Erysiphe necator) fungicide which shows protectant and post-infective activities.
Specifikationer
Kemiskt namn eller material Meptyldinocap
Fysisk form Oil
CAS 131-72-6
Färg Yellow
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H24N2O6
För användning med (applikation) Neuroscience-Neuromodulation
Synonym 2,4-DNOPC
Löslighetsinformation DMSO : ≥ 150 mg/mL (411.65 mM) ∣Ethanol : 100 mg/mL (274.43 mM; Need ultrasonic)
LEDER C/C=C/C(OC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1C(C)CCCCCC)=O
Molekylvikt (g/mol) 364.39
Formel vikt 364.39
Procent renhet 95.54%