Läs mer
Mequindox, TRC
Produktkod. 30481236
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30481236 500 mg
1 options
Produktkod. 30481236 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM225118500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mequindox
CAS 13297-17-1
Molekylformel C11 H10 N2 O3
Kvantitet 500 mg
Synonym Ethanone, 1-(3-methyl-2-quinoxalinyl)-, N,N'-dioxide, Ketone, methyl 3-methyl-2-quinoxalinyl, N,N'-dioxide (8CI), 1-(3-Methyl-1,4-dioxido-2-quinoxalinyl)ethanone, 2-Acetyl-3-methylquinoxaline 1,4-di-N-oxide, 2-Acetyl-3-methylquinoxaline 1,4-dioxide, 2-Methyl-3-acetylquinoxaline 1,4-dioxide, Maqo, Maquindox, Mequindox
InChI formel InChI=1S/C11H10N2O3/c1-7-11(8(2)14)13(16)10-6-4-3-5-9(10)12(7)15/h3-6H,1-2H3
LEDER CC(=O)c1c(C)[n+]([O-])c2ccccc2[n+]1[O-]
IUPAC-namn 1-(3-methyl-1,4-dioxidoquinoxaline-1,4-diium-2-yl)ethanone
Molekylvikt (g/mol) 218.21
Formel vikt 218.0691