Läs mer
Mequitazine Sulfoxide, Mikromol™
Produktkod. 30051558
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051558 25 mg
17933256 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051558 Leverantör Mikromol Leverantörsnummer MM3131.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Mequitazine Sulfoxide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mequitazine Sulfoxide
CAS 40765-34-2
Analyt- eller komponentnamn Mequitazine Sulfoxide
Frakt skick Room Temperature
Molekylformel C20 H22 N2 O S
Certifikat och efterlevnad ISO 17025
Synonym 10-(1-Azabicyclo[2.2.2]oct-3-ylmethyl)-10H-phenothiazine 5-Oxide, Mequitazine S-oxide, Mequitazine Sulfoxide
InChI formel InChI=1S/C20H22N2OS/c23-24-19-7-3-1-5-17(19)22(18-6-2-4-8-20(18)24)14-16-13-21-11-9-15(16)10-12-21/h1-8,15-16H,9-14H2
LEDER O=S1c2ccccc2N(CC3CN4CCC3CC4)c5ccccc15
IUPAC-namn 10-(1-azabicyclo[2.2.2]octan-3-ylmethyl)phenothiazine 5-oxide
Molekylvikt (g/mol) 338.47
Kvantitet 25 mg
Formel vikt 338.469 g/mol