Learn More
Ansell™ Mercury™ 43-113 Series Yellow Aramid Fiber Heavy Weight Gloves
Product Code. 12669721 Shop All Ansell Products
Change view
Click to view available options
Size:
10
11
Unit Size:
6 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12669721 10 6 pairs
12659721 11 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12669721 Supplier Ansell™ Supplier No. 43113100
  • ONLINE EXCLUSIVE PRICE
    4505.00 SEK
    2725.48 SEK / 6 par
    Save 1779.52 SEK
    39% Av
Beräknad leverans: 02-12-2026
Mercury™ 43-113 Series Yellow Aramid Top of the range gloves for the most exacting hot applications. Also provides fantastic resistance to cuts, tears and abrasion.
  • Soft, comfortable and absorbent
  • Designed for intermittent dry heat contact up to 350°C
  • Knitted construction
  • Cotton Kevlar™ Aramid fibers liner
  • Long knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Dupont™ Kevlar™ aramid fibers
Cuff Material Dupont™ Kevlar™ aramid fibers
Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color Yellow
Latex Free Yes
Material Para-Aramid
Product Type Heavyweight Gloves
Size 10
Temperature (English) Max. 660°F
Temperature (Metric) Max 350°C
Glove Cuff Color Yellow
Glove Cuff Style Long Knitwrist
Liner Material Cotton
Lining Lined
Quantity 72 Pairs
Resistant To Heat, cuts, tears and abrasion resistance
Series 43-113