Läs mer
Merethoxylline Procaine, TRC
Produktkod. 30466705
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30466705 25 mg
1 options
Produktkod. 30466705 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22558025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Procaine Merethoxylline
CAS 60064-28-0
Molekylformel C15 H21 Hg N O7 . C13 H20 N2 O2
Kvantitet 25 mg
Synonym Merethoxylline Procaine, Mercurate(1-), [3-[[2-(carboxylatomethoxy)benzoyl-κO]amino]-2-(2-methoxyethoxy)propyl-κC]hydroxy-, hydrogen, compd. with 2-(diethylamino)ethyl 4-aminobenzoate (1:1:1), Mercurate(1-), [3-[[2-(carboxylatomethoxy)benzoyl-κO]amino]-2-(2-methoxyethoxy)propyl-κ
InChI formel InChI=1S/C15H20NO6.C13H20N2O2.Hg.H2O/c1-11(21-8-7-20-2)9-16-15(19)12-5-3-4-6-13(12)22-10-14(17)18;1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;;/h3-6,11H,1,7-10H2,2H3,(H,16,19)(H,17,18);5-8H,3-4,9-10,14H2,1-2H3;;1H2/q;;+1;/p-1
LEDER CCN(CC)CCOC(=O)c1ccc(N)cc1.COCCOC(CNC(=O)c2ccccc2OCC(=O)O)C[Hg]O
IUPAC-namn [3-[[2-(carboxymethoxy)benzoyl]amino]-2-(2-methoxyethoxy)propyl]-hydroxymercury;2-(diethylamino)ethyl 4-aminobenzoate
Molekylvikt (g/mol) 764.23
Formel vikt 765.255