Läs mer
Merimepodib, MedChemExpress
Produktkod. 16495948
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16495948 10 mg -
16482097 50 mg -
16478577 5 mg -
16406386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16495948 Leverantör Medchem Express Leverantörsnummer HY13986
MedChemExpress Merimepodib (VX-497) is a noncompetitive and oral inhibitor of inosine monophosphate dehydrogenase (IMPDH) with broad spectrum antiviral activities.
Merimepodib (VX-497) is a noncompetitive and oral inhibitor of inosine monophosphate dehydrogenase (IMPDH) with broad spectrum antiviral activities.
Specifikationer
Kemiskt namn eller material Merimepodib
Fysisk form Solid
CAS 198821-22-6
Färg Off-White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C23H24N4O6
För användning med (applikation) COVID-19-anti-virus
Synonym VX-497 MMPD
Löslighetsinformation DMSO : ≥ 31 mg/mL (68.51 mM)
LEDER O=C(O[C@@H]1COCC1)NCC2=CC=CC(NC(NC3=CC=C(C4=CN=CO4)C(OC)=C3)=O)=C2
Molekylvikt (g/mol) 452.46
Formel vikt 452.46
Procent renhet 98.55%