missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERTK Monoclonal Antibody (DS5MMER), APC-eFluor™ 780, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1850.00 SEK - 4605.00 SEK
Specifications
Antigen | MERTK |
---|---|
Clone | DS5MMER |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16616374
|
Affymetrix eBioscience
47-5751-80 |
25 μg |
1850.00 SEK
Pack of 2 |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16683632
|
Affymetrix eBioscience
47-5751-82 |
100 μg |
4605.00 SEK
100µg |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16626374
|
Affymetrix eBioscience
47-5751-82 |
100 μg |
4605.00 SEK
Each |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MERTK Monoclonal antibody specifically detects MERTK in Mouse samples. It is validated for Flow Cytometry.Specifications
MERTK | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
c-Eyk; c-mer; c-mer proto-oncogene tyrosine kinase; Eyk; MER; MER proto-oncogene, tyrosine kinase; MER receptor tyrosine kinase; MERTK; MGC133349; nmf12; Nyk; Proto-oncogene c-Mer; proto-oncogene tyrosine-protein kinase MER; rdy; receptor tyrosine kinase MerTK; Receptor tyrosine tinase gene probably the gene for Rdy; retinal dystrophy; RP38; sMER; sMERTK; soluble MER; soluble MERTK; STK kinase; Tyro 12; Tyro12; Tyrosine-protein kinase Mer | |
Mertk | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
DS5MMER | |
Flow Cytometry | |
APC-eFluor 780 | |
Rat | |
Mouse | |
Q60805 | |
17289 | |
FC-tagged full protein expressed in insect cells | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Mertk |