Learn More
meso-1,2-Diphenyl-1,2-ethanediol, Sigma-Aldrich™
Product Code. 32237775
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
32237775 1 g 1g
1 options
Product Code. 32237775 Supplier Sigma Aldrich Supplier No. 8205260001
 Restricted Item
Beräknad leverans: 26-11-2026
Add to basket
Add to basket
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material meso-1,2-Diphenyl-1,2-ethanediol
CAS 492-70-6
Melting Point 139°C
Molecular Formula HOCH(C6H5)CH(C6H5)OH
Quantity 1 g
Synonym meso-1,2-Diphenyl-1,2-ethanediol, meso-Hydrobenzoin, meso-1,2-Diphenylethylene glycol, meso-1,2-Diphenyl-1,2-ethanediol
Molecular Weight (g/mol) 214.26
Grade Not Available