missing translation for 'onlineSavingsMsg'
Läs mer

meso-Tetra (p-Bromophenyl) Porphine, TRC

Product Code. 30449151
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30449151 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30449151 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT310613500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material meso-Tetra (p-Bromophenyl) Porphine
CAS 29162-73-0
Molekylformel C44H26Br4N4
Kvantitet 500 mg
Synonym 5,10,15,20-Tetrakis(4-bromophenyl)-21H,23H-porphine, 5,10,15,20-Tetra(4-bromophenyl)porphyrin, 5,10,15,20-Tetra(p-bromophenyl)porphyrin, 5,10,15,20-Tetrakis(4-bromophenyl)-21H,23H-porphyrin, 5,10,15,20-Tetrakis(4-bromophenyl)porphyrin, 5,10,15,20-Tetrakis(p-bromophenyl)porphyrin, Tetra(4-bromophenyl)porphyrin, Tetra(p-bromophenyl)porphyrin, Tetrakis(4-bromophenyl)porphyrin, meso-Tetra(4-bromophenyl)porphyrin, meso-Tetra(p-bromophenyl)porphyrin, meso-Tetrakis(4-bromophenyl)porphyrin
InChI formel InChI=1S/C44H26Br4N4/c45-29-9-1-25(2-10-29)41-33-17-19-35(49-33)42(26-3-11-30(46)12-4-26)37-21-23-39(51-37)44(28-7-15-32(48)16-8-28)40-24-22-38(52-40)43(36-20-18-34(41)50-36)27-5-13-31(47)14-6-27/h1-24,49,52H/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-
LEDER Brc1ccc(cc1)c2c3C=Cc([nH]3)c(c4ccc(Br)cc4)c5ccc(n5)c(c6ccc(Br)cc6)c7ccc([nH]7)c(c8ccc(Br)cc8)c9ccc2n9
IUPAC-namn (5Z,10Z,14Z,19Z)-5,10,15,20-tetrakis(4-bromophenyl)-21,23-dihydroporphyrin
Molekylvikt (g/mol) 930.32
Formel vikt 925.8891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.