Läs mer
Mesosulfuron-methyl, TRC
Produktkod. 30133551
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133551 5 g
17911735 1 g
17962825 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133551 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2252555G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 208465-21-8
Molekylformel C17 H21 N5 O9 S2
Molekylvikt (g/mol) 503.51
Synonym AE-F 130060, AE-F 130060-00, Mesomaxx, Mesosulfuron-methyl, Shima
IUPAC-namn methyl 2-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoyl]-4-(methanesulfonamidomethyl)benzoate
LEDER COC(=O)c1ccc(CNS(=O)(=O)C)cc1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2
Specifikationer
Kemiskt namn eller material Mesosulfuron-methyl
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C17H21N5O9S2/c1-29-13-8-14(30-2)20-16(19-13)21-17(24)22-33(27,28)12-7-10(9-18-32(4,25)26)5-6-11(12)15(23)31-3/h5-8,18H,9H2,1-4H3,(H2,19,20,21,22,24)
Formel vikt 503.0781
Procent renhet >95%