Läs mer
Mesosulfuron, TRC
Produktkod. 30472598
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472598 100 mg
30484120 10 mg
30468890 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472598 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM314120100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 400852-66-6
Molekylformel C16 H19 N5 O9 S2
Molekylvikt (g/mol) 489.4802
Synonym 2-[[[[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl]amino]sulfonyl]-4-[[(methylsulfonyl)amino]methyl]benzoic acid, AE-F 154851, Mesosulfuron
IUPAC-namn 2-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoyl]-4-(methanesulfonamidomethyl)benzoic acid
LEDER COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2cc(CNS(=O)(=O)C)ccc2C(=O)O)n1
Specifikationer
Kemiskt namn eller material Mesosulfuron
Kvantitet 100 mg
InChI formel InChI=1S/C16H19N5O9S2/c1-29-12-7-13(30-2)19-15(18-12)20-16(24)21-32(27,28)11-6-9(8-17-31(3,25)26)4-5-10(11)14(22)23/h4-7,17H,8H2,1-3H3,(H,22,23)(H2,18,19,20,21,24)
Formel vikt 489.0624