Läs mer
meta-Methoxytopolin Riboside, TRC
Produktkod. 30473338
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473338 100 mg
30478599 25 mg
30480133 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30473338 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM271665100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 101565-95-1
Molekylformel C18H21N5O5
Molekylvikt (g/mol) 387.39
Synonym N-[(3-Methoxyphenyl)methyl]adenosine, MemTR, 6-(3-Methoxybenzylamino)-9-ß-dribofuranosylpurine, N-[(3-Methoxyphenyl)methyl]-adenosine
IUPAC-namn (2R,3R,4S,5R)-2-(hydroxymethyl)-5-[6-[(3-methoxyphenyl)methylamino]purin-9-yl]oxolane-3,4-diol
LEDER COc1cccc(CNc2ncnc3c2ncn3[C@@H]4O[C@H](CO)[C@H](O)[C@@H]4O)c1
Specifikationer
Kemiskt namn eller material meta-Methoxytopolin Riboside
Kvantitet 100 mg
InChI formel InChI=1S/C18H21N5O5/c1-27-11-4-2-3-10(5-11)6-19-16-13-17(21-8-20-16)23(9-22-13)18-15(26)14(25)12(7-24)28-18/h2-5,8-9,12,14-15,18,24-26H,6-7H2,1H3,(H,19,20,21)/t12-,14+,15+,18-/m1/s1
Formel vikt 387.1543